QLA Outer Glass with Flat Bottom Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 mL; Clear
Representative image only.
QLA Outer Glass with Flat Bottom Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 mL; Clear
| Mfr: GLA100-VBIO| Item: 4130608
Biodissolution Glassware for automatic testing of dosage forms
₹39,800.00INR / Each
₹39,800.00INR / Each
Product Description
Key Features
  • Flat bottom
More About this Item
Class A non-graduated outer glass cylinder holds inner reciprocating cylinder for biodissolution processes. Cylinder is engineered and manufactured to the precise specifications of USP guidelines. All QLA products are manufactured to ISO9001 quality standards.
.
ISO 9001:2008 Certified Supplier
Specifications
  • Capacity (mL)100
  • MaterialGlass
  • DescriptionOuter Glass with Flat Bottom Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 mL; Clear