QLA Graduated Cylinder, 100 mL, Funnel Top, Hexagonal Base; Class A

Representative image only.

QLA Graduated Cylinder, 100 mL, Funnel Top, Hexagonal Base; Class A

| Mfr: GLA1CA-01| Item: 4130610

QLA Graduated Cylinder designed for tapped density testing

₹46,900.00INR / Each

₹46,900.00INR / Each
Product Description

Key Features

  • Wide funnel top and hexagonal base for easy use Class A calibrated at 20°C

More About this Item

This reusable graduated cylinder is ideal for tapped density sample testing. The wide hexagonal shaped base provides stability while on a benchtop or locked in a testing apparatus and makes the cylinder roll-resistant. The wide funnel top aids in pouring.

.

Specifications
  • Capacity (mL)100
  • MaterialGlass
  • DescriptionGraduated Cylinder, 100 mL, Funnel Top, Hexagonal Base; Class A
Reviews
Q&A