QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, 2 µm, UHMW PE; 100/PK
Representative image only.
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, 2 µm, UHMW PE; 100/PK
| Mfr: FIL002-01-a| Item: 4130885
Achieve reproducibility and consistency from test to test with QLA dissolution filters
₹36,200.00INR / Pkg of 100
₹36,200.00INR / Pkg of 100
Product Description
Key Features
  • Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal
  • 2 µm pore size
  • 100 per pack
  • Made of Ultra High Molecular Weight (UHMW) polyethylene
More About this Item
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds and provide a good flow rate.
.
Specifications
  • MaterialUHMW Polyethylene
  • Pore Size (micron)2
  • Internal Diameter (mm)3.2
  • DescriptionFull-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, 2 µm, UHMW PE; 100/PK