QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, 10 µm, PVDF, Green Micro-Spheres; 1000/PK
Representative image only.
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, 10 µm, PVDF, Green Micro-Spheres; 1000/PK
| Mfr: FIL010-01K| Item: 4130898
Achieve reproducibility and consistency from test to test with QLA dissolution filters
₹380,000.00INR / Pkg of 1000
₹380,000.00INR / Pkg of 1000
Product Description
Key Features
  • Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal
  • 1000 per pack
  • PVDF Material with green specks added for identification
More About this Item
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds and provide a good flow rate.
.
Specifications
  • MaterialPVDF
  • Pore Size (micron)10
  • Internal Diameter (mm)3.2
  • DescriptionFull-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, 10 µm, PVDF, Green Micro-Spheres; 1000/PK