QLA Footed Vessel for Pharmatest, 1000 mL; Clear Plastic

Representative image only.

QLA Footed Vessel for Pharmatest, 1000 mL; Clear Plastic

| Mfr: PLA900-PT| Item: 4130664

QLA vessels provide more precise and consistent dissolution results

₹49,600.00INR / Each

₹49,600.00INR / Each
Product Description

Key Features

  • 1000 mL capacity
  • Footed base for extra stability
  • Clear plastic vessel

More About this Item

Vessels are manufactured using a unique molding process to provide a precise and consistent inside diameter and spherical radius to meet the physical dimensions of the USP.

Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.

.

Specifications
  • Capacity (mL)1000
  • MaterialPlastic
  • Bottle ColorClear
  • Special FeaturesFooted
  • DescriptionFooted Vessel for Pharmatest, 1000 mL; Clear Plastic
Reviews
Q&A