QLA Footed Vessel for Erweka, 1000 mL; Clear Plastic

Representative image only.

QLA Footed Vessel for Erweka, 1000 mL; Clear Plastic

| Mfr: PLA900-EW| Item: 4130662

QLA vessels provide more precise and consistent dissolution results

₹49,600.00INR / Each

₹49,600.00INR / Each
Product Description

Key Features

  • Achieve reproducible results with precision molded vessels
  • Footed base for extra stability
  • Clear plastic vessel
  • 1000 mL capacity

More About this Item

Standard glass vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.

.

Specifications
  • Capacity (mL)1000
  • MaterialPlastic
  • Bottle ColorClear
  • Special FeaturesFooted
  • DescriptionFooted Vessel for Erweka, 1000 mL; Clear Plastic
Reviews
Q&A