QLA Filter Tips Compatible with Agilent and VanKel Dissolution Apparatus, 10 µm, 1/8" ID; 1000/PK
Representative image only.
QLA Filter Tips Compatible with Agilent and VanKel Dissolution Apparatus, 10 µm, 1/8" ID; 1000/PK
| Mfr: FILTIP-10| Item: 4130996
Achieve reproducibility and consistency from test to test with QLA dissolution filters
₹108,000.00INR / Pkg of 1000
₹108,000.00INR / Pkg of 1000
Product Description
Key Features
  • Ideal for process involving large diameter sample probes Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal
  • 10 µm pore size
More About this Item
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filter feature Ultra High Molecular Weight (UHMW) polyethylene membranes and polypropylene housing.
.
Specifications
  • MaterialUHMW polyethylene membranes; PP housing
  • Pore Size (micron)10
  • Internal Diameter (mm)3.2
  • DescriptionFilter Tips Compatible with Agilent and VanKel Dissolution Apparatus, 10 µm, 1/8" ID; 1000/PK