QLA Custom Basket for Sotax Automated System, 28 Mesh; 52% open area
Representative image only.
QLA Custom Basket for Sotax Automated System, 28 Mesh; 52% open area
| Mfr: CUSTBSK-28ST| Item: 4130858
QLA Baskets dissolution testing with your Sotax apparatus
₹54,000.00INR / Each
₹54,000.00INR / Each
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
  • 28 mesh
  • 52% open area
More About this Item
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Sotax test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Mesh Size28 (52% open area)
  • DescriptionCustom Basket for Sotax Automated System, 28 Mesh; 52% open area