QLA Clip Style Basket for Sotax, 40 Mesh USP, 316 SS; Serialized
Representative image only.
QLA Clip Style Basket for Sotax, 40 Mesh USP, 316 SS; Serialized
| Mfr: BSK040-ST| Item: 4130152
QLA Baskets dissolution testing with your Sotax apparatus
₹37,400.00INR / Each
₹37,400.00INR / Each
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
  • 40 mesh
More About this Item
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Sotax test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Mesh Size40
  • DescriptionClip Style Basket for Sotax, 40 Mesh USP, 316 SS; Serialized