QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 Mesh, 316 SS, Serialized
Representative image only.
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 Mesh, 316 SS, Serialized
| Mfr: BSK100-01| Item: 4130158
QLA Baskets for dissolution testing with your Agilent/VanKel/Varian apparatus
₹142,000.00INR / Each
₹142,000.00INR / Each
Product Description
Key Features
  • Made of 316 electropolished stainless steel
  • All baskets are laser marked with unique serial numbers
  • 100 mesh
More About this Item
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Agilent/VanKel/Varian test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
Not available for sale in the United States.
Specifications
  • Material316 stainless steel
  • Mesh Size100
  • DescriptionClip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 Mesh, 316 SS, Serialized