QLA Bent PEEK Sampling Cannula, Custom, 12"L; 1/8" OD
Representative image only.
QLA Bent PEEK Sampling Cannula, Custom, 12"L; 1/8" OD
| Mfr: CAN012-PV| Item: 4130211
Consistent sampling with QLA Cannula
₹51,900.00INR / Each
₹51,900.00INR / Each
Product Description
Key Features
  • Chemically inert PEEK will not absorb or interfere with active ingredients
  • 12" long
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannula, and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
.
Specifications
  • MaterialPEEK
  • Length (")12
  • DescriptionBent PEEK Sampling Cannula, Custom, 12"L; 1/8" OD