QLA 3 Prong Capsule Sinker; Plastic
Representative image only.
QLA 3 Prong Capsule Sinker; Plastic
| Mfr: CAPWHT-VK| Item: 4130295
Capsule sinkers improve reproducibility
₹11,700.00INR / Each
₹11,700.00INR / Each
Product Description
Key Features
  • Keep floating tablets from sticking to sides of vessels
  • 0.375" (9.5 mm) ID
More About this Item
Sinkers help to ensure tablets dissolve at more reproducible rates and keep them from sticking to the sides of vessels. Select from spring style, basket style, three-prong style, and others.
Specifications
  • MaterialNylon/plastic
  • Diameter (InchShort)3/8
  • Diameter (mm)9.5
  • Description3 Prong Capsule Sinker; Plastic