Pyrex Vista 70024-250 Graduated Glass Cylinder, 250 mL, to deliver, 1/ea

Representative image only.

Pyrex Vista 70024-250 Graduated Glass Cylinder, 250 mL, to deliver, 1/ea

Pyrex Vista | Mfr: 70024-250| Item: 3450474

Calibrated "to deliver"

₹9,100.00INR / Each

for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹9,100.00INR / Each

Buy It Together

Current Item

Pyrex Vista 70024-250 Graduated Glass Cylinder, 250 mL, to deliver, 1/ea
Pyrex Vista 70024-250 Graduated Glass Cylinder, 250 mL, to deliver, 1/ea
3450474

₹9,100.00INR / Each
+ =
Total Price 23600.00 ADD 2 ITEMS TO CART
Product Description

More About this Item

Manufactured of Class A borosilicate glass, these metric scale cylinders are enameled with blue graduations for easy reading. Cylinder capacity is in accordance wtih ASTM E-1272. 10mL size has a funnel top for easy filling.

Specifications
  • MaterialGlass
  • Capacity (mL)250
  • Graduation Interval (mL)2
  • Tolerance (mL)±2.0
  • DescriptionGraduated Glass Cylinder, 250 mL, to deliver, 1/ea
Reviews
Q&A