Pyrex 2157-100TJ Jacketed Concentrator Tube 1-Step Modular Glass System, 100 mL
Representative image only.
Pyrex 2157-100TJ Jacketed Concentrator Tube 1-Step Modular Glass System, 100 mL
Pyrex | Mfr: 2157-100TJ| Item: 3451002
₹95,100.00INR / Case of 1
₹95,100.00INR / Case of 1
Product Description
More About this Item
Accelerated One-Step™ modular extractor/concentrator performs extraction, concentration, and drying—all in one apparatus! Features PTFE hydrophobic membranes for speedy drying. Meets EPA and CLP standards. Includes extractor cup and body, Snyder sidearm, membrane assembly, and connectors. Order concentrator tube and condenser separately below to fit your specific application.
Specifications
  • DescriptionJacketed Concentrator Tube 1-Step Modular Glass System, 100 mL