PTFE-Coated Steel Forceps, Pointed Tips, 3-7/8"L; 2/Pk
Representative image only.
PTFE-Coated Steel Forceps, Pointed Tips, 3-7/8"L; 2/Pk
| Item: 0644000
PTFE coating offers excellent heat and chemical resistance
₹14,800.00INR / Bag of 2
₹14,800.00INR / Bag of 2
Product Description
More About this Item
PTFE-coated for heat and chemical resistance. Coating prevents scratching and marring of delicate surfaces. Nonsticking and noncorroding. Forceps measure 3-7/8"L. Autoclavable.
Specifications
  • Overall Length (in)4
  • Overall Length (cm)10.16
  • MaterialPTFE-coated Stainless Steel
  • SterileYes
  • DescriptionPTFE-Coated Steel Forceps, 3-7/8" L, Pointed Tips; 2/Pk