ProChem PISS 8250 Steam Indicator/integrator - Class 5, 8" Long, 250 Strips/pk
Representative image only.
ProChem PISS 8250 Steam Indicator/integrator - Class 5, 8" Long, 250 Strips/pk
ProChem | Mfr: PISS 8250| Item: 9470221
Call For Price
Call For Price
Product Description
More About this Item
The indicator provides an integrated response to steam sterilization at 121degC or 132degC to 135degC
for pre-vaccum sterilizers.
Specifications
  • Depth (in)8
  • DescriptionSteam Indicator/integrator - Class 5, 8" Long, 250 Strips/pk