PRO Scientific Multi-Gen 7XL Homogenizer Package, 1.5 to 15 mL Tubes; 120 V
Representative image only.
PRO Scientific Multi-Gen 7XL Homogenizer Package, 1.5 to 15 mL Tubes; 120 V
PRO Scientific | Mfr: PK-01200PMGXL| Item: 0475103
Avoid cross-contamination while homogenizing multiple samples
₹747,000.00INR / Each
₹747,000.00INR / Each
Product Description
Includes
Bio-Gen PRO200® homogenizer, 12 generator probes, adapter, carrying case, and power cord.
Key Features
  • 12-Pack of Reusable Stainless Steel Probes, processing in 1.5ml-15ml tubes
  • Environmentally friendly, long life span, and economical option
  • Open slotted tooth design- ideal for soft to tough samples
  • Made in the USA
  • 2 year warranty
More About this Item
The Multi-Gen 7XL homogenizing kit is ideal for processing multiple samples directly inside 1.5 to 15 mL tubes without the risk of cross-contamination or need to clean probes in between samples. Environmentally friendly option over plastic disposable probes with a long life span and economical operation within the first year. Included homogenizer has a 2-year warranty with a 144-watt, high-torque motor that features a variable-speed adjustment, allowing homogenization in seconds. A separate on/off switch located at the top of the unit provides easy access and increased safety. The included 12-Pack of Multi-Gen 7XL probes have a 7mm X 115m open-slotted tooth design, constructed of 316 stainless steel and PTFE. They can be reused an unlimited number of times, process difficult samples like tough tissues, and can be sterilized by any method, including flaming and autoclaving. An adapter allows the homogenizer to connect a clean probe and disconnect a used one quickly and easily, avoiding the chance of accidental contamination.
UL Listed.UL Listed to Canadian safety standards.Two year warranty
Specifications
  • DescriptionHomogenizer Package, 1.5 to 15 mL Tubes; 120 V