Plas-Labs 800-PRV Glove Box Pressure Relief Valve, 1/8" NPT Stainless Steel
Representative image only.
Plas-Labs 800-PRV Glove Box Pressure Relief Valve, 1/8" NPT Stainless Steel
Plas-Labs | Mfr: 800-PRV| Item: 3474968
Eliminates unwanted positive pressure build-up
₹46,000.00INR / Each
₹46,000.00INR / Each
Product Description
Key Features
  • Directs exhausted air away from operators
  • 1 year warranty
More About this Item
Useful for eliminating unwanted positive pressure build-up in glove boxes. Acrylic relief valve features 1/8" NPT stainless steel inlet and outlet ports. Exhausted air can be directed away from operators by a hose connection to the outlet port.
1 Year Warranty
Specifications
  • Outlet Port1/8" NPT(F)
  • Connection TypeNPT(M) x NPT(F)
  • Inlet Port1/8" NPT(M)
  • Width (in)1
  • Height (in)2
  • Length (in)1
  • DescriptionGlove Box Pressure Relief Valve, 1/8" NPT Stainless Steel