Plas-Labs 800-PC Palladium canister, filled with ¾-lb palladium pellets for catalyst heater.
Representative image only.
Plas-Labs 800-PC Palladium canister, filled with ¾-lb palladium pellets for catalyst heater.
Plas-Labs | Mfr: 800-PC| Item: 3474964
Optimize performance of your Plas-Labs glove boxes
Call For Price
Call For Price
Product Description
Key Features
  • 1 year warranty
More About this Item
Stainless steel support carts feature non-scuff casters with brakes and locks.
The Palladium canister is filled with 3⁄4-lb palladium pellets for use with catalyst heater models 34749-56 and -58.
1 Year Warranty
Specifications
  • DescriptionPalladium canister, filled with ¾-lb palladium pellets for catalyst heater.