Phipps & Bird Replacement B-KERbeaker
Representative image only.
Phipps & Bird Replacement B-KERbeaker
| Mfr: 7790-630| Item: 9952050
Optimize water analysis
₹31,500.00INR / Each
₹31,500.00INR / Each
Buy It Together
Current Item
Phipps & Bird Replacement B-KERbeaker
Phipps & Bird Replacement B-KERbeaker
9952050
₹31,500.00INR / Each
+ =
Total Price 31500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Electrodes and probes, carrying cases, cables, calibration standards, and replacement parts.
The laboratory B-Ker Jar is molded of strong durable 1/4" thick acrylic. Compatible with most Phipps & Bird two- four- and six-paddle Jartesters. Includes sampling valve. 10cm setting distance, 1/4" wall thickness,
1 Year Warranty
Specifications
  • Compatible WithPhipps & Bird paddle stirrer
  • DescriptionReplacement B-KERbeaker