Parker Hannifin Zero Air Gas Generator Maintenance Kit for 75-83 Model Gas Generators
Representative image only.
Parker Hannifin Zero Air Gas Generator Maintenance Kit for 75-83 Model Gas Generators
Parker Hannifin | Mfr: MK7583 | Item: 8644253
Optimize performance of your Parker Balston Zero Air Generator
Call For Price
Call For Price
Product Description
Includes
one first stage coalescing prefilter cartridge (P/N 100-12-DX), one second stage coalescing filter cartridge (P/N 100-12-BX), and one Grade GS final membrane filter (P/N GS-050-05-95).
Key Features
  • Installation and maintenance parts for your Parker Balston Zero Air Generators
  • 1 year warranty
More About this Item
Improve operation of your Parker Balston Zero Air Generators with maintenance and installation kits.
Installation kit includes all the fittings required to bring generator online quickly and easily.
Maintenance kits provide a convenient, a one-year supply of replacement consumables need to keep your generator running smoothly.
Conformite Europeenne1 Year Warranty
Specifications
  • Compatible WithParker Balston 75-83 Model Gas Generators
  • DescriptionZero Air Gas Generator Maintenance Kit for 75-83 Model Gas Generators