Parker Hannifin 5/050-05-BX Replacement Compressed-Air Filter; 6.0cfm; Port Size; 1/4" NPT(F); 5/Pk
Representative image only.
Parker Hannifin 5/050-05-BX Replacement Compressed-Air Filter; 6.0cfm; Port Size; 1/4" NPT(F); 5/Pk
Parker Hannifin | Mfr: 5/050-05-BX | Item: 0291751
Removes oil, water, and solid contaminants
₹39,100.00INR / Box of 5
₹39,100.00INR / Box of 5
Product Description
Key Features
  • No need for special tools
More About this Item
These replacement filters are ideal for Parker Hannifin compressed air filters. Excellent in removing oil, water, and contaminants.
Specifications
  • Compatible WithParker Hannifin 8A02N-OB2-BX In-Line Compressed Air Filter (02917-00)
  • DescriptionReplacement Compressed-air filter; 6.0cfm; port size; 1/4" NPT(F); 5/box