Pacer 10195 5001 EXTENSION ROD FOR DA AN
Representative image only.
Pacer 10195 5001 EXTENSION ROD FOR DA AN
Pacer | Mfr: 10195| Item: 2609123
Offers robust metal probes, easy to use instruments, and highly accurate measurements down to ultra-low flow rates
₹4,400.00INR / Each
₹4,400.00INR / Each
Product Description
Includes
Hard carrying case, 3 alkaline batteries, set of three rigid extension rods with handle, flexible extension rod, operation manual, 2.75 inch rotating vane probe and 5 foot probe cable.
Key Features
  • Industrial grade instrument
  • Rugged 2.75 inch diameter metal probe
  • Air Speed display in ft/min or m/s
  • Min / Max capture
  • Data Hold and Auto Power Off
  • Variety of sample rates for flow averaging
  • Large display with Backlight
  • Includes NIST-Traceable calibration certificate
More About this Item
Pacer’s Premier Series of Rotating Vane Anemometers offers robust metal probes, easy to use instruments, and highly accurate measurements down to ultra-low flow rates. Models are available for automatically calculating volumetric flow rates or for measuring both air velocity and temperature. Choose from many popular options such as Analog Voltage output and USB data output. Applications include HVAC, industrial process control, cleanroom monitoring, laboratory, research, and aerospace development.