Optical Dissolved Oxygen Transmitter, submersible, 4-20 mA output
Representative image only.
Optical Dissolved Oxygen Transmitter, submersible, 4-20 mA output
| Item: 5320122
Submersible 2-wire 4 to 20 mA transmitter for long-term monitoring applications
₹439,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹439,000.00INR / Each
Buy It Together
Current Item
Optical Dissolved Oxygen Transmitter, submersible, 4-20 mA output
Optical Dissolved Oxygen Transmitter, submersible, 4-20 mA output
5320122
₹439,000.00INR / Each
+ =
Total Price 463700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Optical technology provides long-term stability
  • Reduced maintenance—no need to replace membranes or electrolyte
  • Autoclean nozzle allows sensor cleaning via external air pressure
  • 3 year warranty
More About this Item
Optical dissolved oxygen transmitters offer a variety of benefits of other sensor types. Sensor uses fluorescence to measure dissolved oxygen eliminating the need for electrolyte or membranes, and reducing overall maintenance costs. Highly stable optical technology reduces the need for recalibration making these transmitters excellent for long-term monitoring applications. Optical sensors are also flow insensitive, which means readings are unaffected by water velocity.
Two-wire design simplifies installation. 4 to 20 mA output signal can be directly connected to a chart recorder or datalogger—ideal for remote monitoring applications. Optional universal input display/controller (order separately) provides a local display for measured readings. Use optical transmitters for groundwater monitoring, industrial effluent analysis, thermocline profiling, wastewater, aquaculture, and more!
3 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier
Specifications
  • Meter TypeTransmitter
  • Min Dissolved Oxygen0 ppm
  • Max Dissolved Oxygen20 ppm
  • Temperature compensationAutomatic
  • Output4 to 20 mA
  • DescriptionOptical Dissolved Oxygen Transmitter, submersible, 4-20 mA output