Optical Dissolved Oxygen Sensor for 53201-10
Representative image only.
Optical Dissolved Oxygen Sensor for 53201-10
| Item: 5320112
Optimize water analysis
₹336,000.00INR / Each
₹336,000.00INR / Each
Buy It Together
Current Item
Optical Dissolved Oxygen Sensor for 53201-10
Optical Dissolved Oxygen Sensor for 53201-10
5320112
₹336,000.00INR / Each
+ =
Total Price 336000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
  • 3 year warranty
More About this Item
Electrodes and probes, carrying cases, cables, calibration standards, and replacement parts.
This unique submersible probe has been designed to measure dissolved oxygen based on fluorescent technology. The measuring system consists of an optical device complete with a layer of fluorescent material, electronic circuit with an exciting beam for the fluorescence detection, built-in amplifier, Pt1000 for temperature compensation, digital input for calibration and configuration, and a nozzle for the autoclean by external pressure air. The probe is powered by controler 53201-10. Most common applications include water quality monitoring, municipal water treatment, and industrial water treatment.
3 Year WarrantyConformite Europeenne
Specifications
  • Compatible WithOptical DO controller, 53201-10
  • DescriptionOptical Dissolved Oxygen Sensor for 53201-10