Ohaus Vortex-Genie SI-BG05 Disruptor Disruptor Beads, 0.5 mm Diameter; 8 oz Bottle

Representative image only.

Ohaus Vortex-Genie SI-BG05 Disruptor Disruptor Beads, 0.5 mm Diameter; 8 oz Bottle

Ohaus | Mfr: SI-BG05| Item: 0472491

₹9,200.00INR / Each

₹9,200.00INR / Each

Buy It Together

Current Item

Ohaus Vortex-Genie SI-BG05 Disruptor Disruptor Beads, 0.5 mm Diameter; 8 oz Bottle
Ohaus Vortex-Genie SI-BG05 Disruptor Disruptor Beads, 0.5 mm Diameter; 8 oz Bottle
0472491

₹9,200.00INR / Each
+
Ohaus Stainless Steel Beads, 5 mm; 100/PK
5140217

₹15,100.00INR / Each
=
Total Price 24300.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Compatible with Disruptor Genie and Digital Disruptor Genie.

As of September 2025 Scientific Industries sold the Vortex Genie product line to Ohaus

Specifications
  • Compatible WithDisruptor Genie
  • DescriptionDisruptor Disruptor Beads, 0.5 mm Diameter; 8 oz Bottle
Reviews
Q&A