Ohaus Universal Platform, Stainless Steel, 46 x 46 cm, for use with Incubating Shakers

Representative image only.

Ohaus Universal Platform, Stainless Steel, 46 x 46 cm, for use with Incubating Shakers

Ohaus | Mfr: 30400055| Item: 5302788

Expand your incubating shaker capabilities

₹113,000.00INR / Each

₹113,000.00INR / Each
Product Description

Key Features

  • 2 year warranty

More About this Item

Increase the amount of usable space for your samples by adding an universal platform.

Two year warranty
Specifications
  • MaterialStainless Steel
  • Compatible WithOhaus incubating shakers
  • Length (InchShort)18
  • Width (InchShort)18
  • Width (cm)45.72
  • Length (cm)45.72
  • DescriptionUniversal Platform, Stainless Steel, 46 x 46 cm, for use with Ohaus Incubating Shakers
  • Warranty2 year warranty
Reviews
Q&A