Ohaus Sticky Mat, 22x18 cm, for use with Shakers
Representative image only.
Ohaus Sticky Mat, 22x18 cm, for use with Shakers
Ohaus | Mfr: 30770939| Item: 5302751
Expand your orbital digital shaker capabilities
₹20,100.00INR / Each
₹20,100.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Optimize your orbital digital shaker with the following accessories. Choose from a variety of platforms and matts to to safely secure your samples. A variety of platform options are available to hold irregular samples or traditional flask clamps, media bottle clamps, and tube racks. Platforms can increase the amount of usable space for your samples, and in some cases can be stacked to increase throughput and double the capacity.
Please note, the sticky matt should not exceed the maximum speed of 250 rpm.. Application temperature not to exceed 65 °C (149 °F). To restore adhesive quality, clean with water and air dry.
1 Year Warranty
Specifications
  • Compatible WithEndeavor 500 E51LD0403 and E51LD0420 Orbital Shakers
  • Length (InchShort)7
  • Width (InchShort)8 3/4
  • Width (cm)22
  • Length (cm)18
  • DescriptionMat Sticky, 22x18 cm, for use with Ohaus Shakers
  • Warranty1 year warranty