Ohaus Standard-Duty Incubating Orbital Microplate Shaker, 3 mm Orbit

Representative image only.

Ohaus Standard-Duty Incubating Orbital Microplate Shaker, 3 mm Orbit

Ohaus | Mfr: ISLDMPHDG| Item: 5302783

Incubator and shaker in one unit saves valuable benchtop space

₹880,000.00INR / Each

₹880,000.00INR / Each

Buy It Together

Current Item

Ohaus Standard-Duty Incubating Orbital Microplate Shaker, 3 mm Orbit
Ohaus Standard-Duty Incubating Orbital Microplate Shaker, 3 mm Orbit
5302783

₹880,000.00INR / Each
+
Ohaus Micro Tube Rack for use with Shaker Platforms
5302711

₹48,800.00INR / Each
=
Total Price 928800.00 ADD 2 ITEMS TO CART
Product Description

Includes

Aluminum tray and detachable three-wire cord with plug.

Key Features

  • Control and view speed, temperature, and time independently
  • 3 mm (0.1”) orbital motion
  • Triple eccentric drive systems provide stable orbital motion
  • Microprocessor-control offers accurate shaking speed
  • View actual/setpoint temperatures
  • Count up/count down timer
  • Holds up to four microplates or deepwell plates, or two micro tube racks
  • 2 year warranty

More About this Item

These standard-duty incubating orbital shakers are perfect for immunoassay, hybridization, PCR, cell cultures, and enzyme reaction applications. Independent controls for speed, temperature, and time and an intuitive touchpad make it easy to set and view the parameters. Digital speed control offers reliable and repeatable shaking speed. Heat samples up to 65 °C; a dome cover helps maintain uniform chamber temperature. Built with operator safety in mind, a “Caution Hot” indicator illuminates if the chamber air temperature is above 40 °C (104 °F). Set the timer to display accumulated time (continuous mode) or remaining time (timed mode) from 1 second to 9999 minutes. Use the included aluminum tray to hold clamps, trays, tacks, or plates for secure shaking.

Conformite EuropeenneUL Listed.Canadian Standards AssociationTwo year warranty
Specifications
  • Orbit Size (in)0.1
  • Orbit Size (mm)3
  • Min Speed (rpm)100
  • Max Speed (rpm)1200
  • Speed Accuracy (rpm)±2% of set speed up to 999 rpm, ±5% from 1000 to 1200 rpm
  • Capacity4 microplates or 2 micro-tube racks
  • Width (in)11
  • Height (in)7.75
  • Depth (in)17
  • Width (cm)27.9
  • Height (cm)19.7
  • Depth (cm)43.2
  • Max Operating Temperature (° C)65
  • Temperature Uniformity±0.5 °C at 37 °C
  • TimerYes
  • Timer Range1 second to 9999 minutes
  • DisplayThree independent LEDs
  • Housing MaterialCast aluminum
  • Power (VAC)120
  • Power (Hz)50/60
  • Power (amps)5
  • DescriptionStandard-Duty Incubating Orbital Microplate Shaker, 3 mm Orbit
  • Warranty2 years
Reviews
Q&A