Ohaus SI-1613 Flask Clamp for Orbital-Genie, 500 mL, 1/ea

Representative image only.

Ohaus SI-1613 Flask Clamp for Orbital-Genie, 500 mL, 1/ea

Ohaus | Mfr: SI-1613| Item: 5182104

Enhance the versatility of your Scientific Industries Orbital Shaker

₹12,600.00INR / Each

₹12,600.00INR / Each
Product Description

More About this Item

Choose from a wide range of interchangeable platforms, a stacking try, or clamps to increase or change the capacity of your shaker.

Orbital-Genie platform accepts 16 clamps of this size, Mini 300 Orbital-Genie accepts 9. Mix and match clamp sizes.

As of September 2025 Scientific Industries sold the Vortex Genie product line to Ohaus

Specifications
  • Capacity (mL)500
  • Compatible WithScientific Industries Genie Temp-Shaker with flask clamp platform 51603-38
  • DescriptionFlask Clamp for Orbital-Genie, 500 mL, 1/ea
Reviews
Q&A