Ohaus Large Vessel Holder, 500 mL/1 L Flask for Vortex Mixers

Representative image only.

Ohaus Large Vessel Holder, 500 mL/1 L Flask for Vortex Mixers

Ohaus | Mfr: 30400211| Item: 5302770

Enhance your vortex mixing options

₹15,200.00INR / Each

₹15,200.00INR / Each

Buy It Together

Current Item

Ohaus Large Vessel Holder, 500 mL/1 L Flask for Vortex Mixers
Ohaus Large Vessel Holder, 500 mL/1 L Flask for Vortex Mixers
5302770

₹15,200.00INR / Each
+
Ohaus Replacement Universal Holder for Vortex Mixers
5302779

₹18,700.00INR / Each
=
Total Price 33900.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Holds 500 mL and 1 L flasks

More About this Item

A wide range of accessories are available for your vortex mixers, allowing you to enhance your throughput by mixing multiple samples simultaneously. You can effortlessly mix up to 52 microtubes or eight microplates at once. Simply choose the holder that fits your sample size, or opt for the flat insert and customize it to meet your specific needs.

Maximize your efficiency in the lab with the addition of a universal holder, microplate holder, vessel holder, or foam insert!

Please note: Requires universal holder (model 53027-79).

1 Year Warranty
Specifications
  • MaterialPlastic
  • Compatible WithOhaus heavy-duty vortex mixers
  • DescriptionLarge Vessel Holder, 500 mL/1 L Flask for Vortex Mixers
  • Warranty1 year
Reviews
Q&A