Ohaus i-C71 Roller Top, 18 x 18", for Courier™ 7000 Series Shipping Scales

Representative image only.

Ohaus i-C71 Roller Top, 18 x 18", for Courier™ 7000 Series Shipping Scales

Ohaus | Mfr: 30819649| Item: 0100454

Improve the versatility of your shipping scales

₹88,600.00INR / Each

₹88,600.00INR / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Enhance the performance of your Ohaus Courier shipping scale by adding one or more of the following accessories.

1 Year Warranty
Specifications
  • Compatible WithOhaus Courier™ 7000 Series Shipping Scales
  • Width (InchShort)18
  • Depth (InchShort)18
  • Width (cm)45.7
  • Depth (cm)45.7
  • DescriptionRoller Top, 18 x 18", for Courier™ 7000 Series Shipping Scales
  • Warranty1 year
Reviews
Q&A