Ohaus Genie SI-1617 Adhering Mat, Blue, 18" x 18" for Orbital- Shaker
Representative image only.
Ohaus Genie SI-1617 Adhering Mat, Blue, 18" x 18" for Orbital- Shaker
Ohaus | Mfr: SI-1617| Item: 5182108
Enhance the versatility of your Scientific Industries Orbital Shaker
₹33,400.00INR / Each
₹33,400.00INR / Each
Product Description
More About this Item
Choose from a wide range of interchangeable platforms, a stacking try, or clamps to increase or change the capacity of your shaker.
Mat is naturally adhering and perfect for use with open containers. Rinse clean to renew adhering properties.
As of September 2025 Scientific Industries sold the Vortex Genie product line to Ohaus
Specifications
  • Compatible WithScientific Industries Orbital Shakers
  • Length (InchShort)18
  • Width (InchShort)18
  • DescriptionAdhering Mat, Blue, 18" x 18" for Orbital- Shaker