Ohaus Genie SI-1170 Roto-Shake Stackable Tray
Representative image only.
Ohaus Genie SI-1170 Roto-Shake Stackable Tray
Ohaus | Mfr: SI-1170| Item: 5160386
Expand the capabilities of your shaker
₹42,500.00INR / Each
₹42,500.00INR / Each
Product Description
Key Features
  • Stackable tray increases throughput
More About this Item
A tall non-skid metal tray that can be stacked onto the Roto-Shake Genie's existing tray to rock various containers including T-flasks, petri dishes, boxes, and sealed bags, increasing the capacity for rocking by double-stacking two trays.
As of September 2025 Scientific Industries sold the Vortex Genie product line to Ohaus
Specifications
  • Compatible WithRoto-Shake Genie
  • Length (InchShort)12
  • Width (InchShort)12
  • DescriptionRoto-Shake Stackable Tray