Ohaus Four Place Stackable Microplate Holder for Vortex Mixers
Representative image only.
Ohaus Four Place Stackable Microplate Holder for Vortex Mixers
Ohaus | Mfr: 30400212| Item: 5302765
Enhance your vortex mixing options
₹38,500.00INR / Each
₹38,500.00INR / Each
Buy It Together
Current Item
Ohaus Four Place Stackable Microplate Holder for Vortex Mixers
Ohaus Four Place Stackable Microplate Holder for Vortex Mixers
5302765
₹38,500.00INR / Each
+
Ohaus Four Place Microplate Holder for Vortex Mixers
5302767
₹41,500.00INR / Each
=
Total Price 80000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Hold up to 8 microplates by stacking tray on top of four plate holder
More About this Item
A wide range of accessories are available for your vortex mixers, allowing you to enhance your throughput by mixing multiple samples simultaneously. You can effortlessly mix up to 52 microtubes or eight microplates at once. Simply choose the holder that fits your sample size, or opt for the flat insert and customize it to meet your specific needs.
Maximize your efficiency in the lab with the addition of a universal holder, microplate holder, vessel holder, or foam insert!
Please note: The four place microplate holder (model 53027-67) must be purchased in order to use this four place stackable microplate holder.
1 Year Warranty
Specifications
  • MaterialPlastic
  • Compatible WithOhaus heavy-duty vortex mixers
  • DescriptionFour Place Stackable Microplate Holder for Vortex Mixers