Ohaus Foam Insert, 13 x 22-25 mm Tubes for Vortex Mixers
Representative image only.
Ohaus Foam Insert, 13 x 22-25 mm Tubes for Vortex Mixers
Ohaus | Mfr: 30400223| Item: 5302776
Enhance your vortex mixing options
₹13,100.00INR / Each
₹13,100.00INR / Each
Buy It Together
Current Item
Ohaus Foam Insert, 13 x 22-25 mm Tubes for Vortex Mixers
Ohaus Foam Insert, 13 x 22-25 mm Tubes for Vortex Mixers
5302776
₹13,100.00INR / Each
+
Ohaus Replacement Universal Holder for Vortex Mixers
5302779
₹18,700.00INR / Each
=
Total Price 31800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Holds 22 to 52 mL microtubes
More About this Item
A wide range of accessories are available for your vortex mixers, allowing you to enhance your throughput by mixing multiple samples simultaneously. You can effortlessly mix up to 52 microtubes or eight microplates at once. Simply choose the holder that fits your sample size, or opt for the flat insert and customize it to meet your specific needs.
Maximize your efficiency in the lab with the addition of a universal holder, microplate holder, vessel holder, or foam insert!
Please note: Requires universal holder (model 53027-79).
1 Year Warranty
Specifications
  • MaterialFoam
  • Compatible WithOhaus heavy-duty vortex mixers
  • DescriptionFoam Insert, 13 x 22-25 mm Tubes for Vortex Mixers