Ohaus 83034024 Sinker, Glass, Liquid, Density Kit for Balances
Representative image only.
Ohaus 83034024 Sinker, Glass, Liquid, Density Kit for Balances
Ohaus | Mfr: 83034024| Item: 2060566
₹277,000.00INR / Each
₹277,000.00INR / Each
Product Description
Key Features
  • Use for liquid density determination
  • 1 year warranty
More About this Item
Use your balance to determine density. Dedicated density determination kits make it easy.
Measure liquid density quickly and accurately with this glass sinker density determination kit, designed for seamless use with Ohaus balances. Utilizing the proven sinker method, the kit calculates density based on buoyancy differences, delivering reliable and repeatable results across a wide range of liquids. Ideal for laboratory, quality control, and research environments, this accessory enables efficient and dependable liquid density determination without the need for separate instrumentation. For use with Adventurer, Pioneer, PR Series, Explorer, and Explorer Plus models. Refer to balance manual for compatibility.
1 Year Warranty
Specifications
  • DescriptionSinker, Glass, Liquid, Density Kit for Ohaus Balances