Ohaus 707-00 Metric Attachment Weight Set; 2x1000g, 1x500g
Representative image only.
Ohaus 707-00 Metric Attachment Weight Set; 2x1000g, 1x500g
Ohaus | Mfr: 707-00| Item: 0101401
Easily increase capacity of your triple beam mechanical balance
₹27,900.00INR / Each
₹27,900.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
This three piece weight set consists of two 1,000 g masses, and one 500 g mass. Use to increase the capacity of your Ohaus Triple Beam mechanical balance from 610 g to 2,610 g.
1 Year Warranty
Specifications
  • Mass Size (g)1000
  • Mass Size (kg)1
  • Mass CertificationStatement of Accuracy
  • DescriptionMetric Attachment Weight Set 2x1000g, 1x500g