Ohaus 31059237 Weighing Kit for Balances
Representative image only.
Ohaus 31059237 Weighing Kit for Balances
Ohaus | Mfr: 31059237| Item: 2060560
₹132,000.00INR / Each
₹132,000.00INR / Each
Product Description
Includes
Centrifuge tubes holder, rotary evaporator (bulb) holder, needle holder, and filter paper holder.
Key Features
  • Includes four different holders
  • 1 year warranty
More About this Item
Versatile kit stabilizes irregular lab vessels for easier weighing. Use with Explorer EXR and Explorer Plus EXP balances.
1 Year Warranty
Specifications
  • Compatible WithExplorer and Explorer Plus Analytical, Precision, and Semi-Micro Balances
  • DescriptionWeighing Kit for Ohaus Balances
  • WarrantyOne year