Ohaus 30696599 Powder-Coated Steel Column Kit for Defender 3000 Bases, 38.6" (98 cm) High

Representative image only.

Ohaus 30696599 Powder-Coated Steel Column Kit for Defender 3000 Bases, 38.6" (98 cm) High

Ohaus | Mfr: 30696599| Item: 1134890

Keep your balance running smoothly with these parts and accessories

₹45,500.00INR / Each

₹45,500.00INR / Each
Product Description

Key Features

  • 1 year warranty
Specifications
  • Materialpowder-coated steel
  • Compatible WithDefender 3000 Bases
  • Height (in)38.6
  • Height (cm)98
  • DescriptionPowder-Coated Steel Column Kit for Defender 3000 Bases, 38.6" (98 cm) High
  • Warranty1 Year
Reviews
Q&A