Ohaus 30696598 Powder-Coated Steel Column Kit for Defender 3000 Bases, 26.7" (68 cm) High

Representative image only.

Ohaus 30696598 Powder-Coated Steel Column Kit for Defender 3000 Bases, 26.7" (68 cm) High

Ohaus | Mfr: 30696598| Item: 1134889

Keep your balance running smoothly with these parts and accessories

₹39,200.00INR / Each

₹39,200.00INR / Each
Product Description

Key Features

  • 1 year warranty
Specifications
  • Materialpowder-coated steel
  • Compatible WithDefender 3000 Bases
  • Height (in)26.7
  • Height (cm)68
  • DescriptionPowder-Coated Steel Column Kit for Defender 3000 Bases, 26.7" (68 cm) High
  • Warranty1 Year
Reviews
Q&A