Ohaus 30696597 Powder-Coated Steel Column Kit for Defender 3000 Bases, 13" (33 cm) High

Representative image only.

Ohaus 30696597 Powder-Coated Steel Column Kit for Defender 3000 Bases, 13" (33 cm) High

Ohaus | Mfr: 30696597| Item: 1134888

Keep your balance running smoothly with these parts and accessories

₹32,700.00INR / Each

₹32,700.00INR / Each
Product Description

Key Features

  • 1 year warranty
Specifications
  • Materialpowder-coated steel
  • Compatible WithDefender 3000 Bases
  • Height (in)13
  • Height (cm)33
  • DescriptionPowder-Coated Steel Column Kit for Defender 3000 Bases, 13" (33 cm) High
  • Warranty1 Year
Reviews
Q&A