Ohaus 30400138 Double Tier Platform Kit, Stainless Steel, 29.8 x 22.2 cm, for use with Waving Shaker SHWV02DG

Representative image only.

Ohaus 30400138 Double Tier Platform Kit, Stainless Steel, 29.8 x 22.2 cm, for use with Waving Shaker SHWV02DG

Ohaus | Mfr: 30400138| Item: 5302755

Optimize your rocking and waving shakers capabilities

₹49,200.00INR / Each

₹49,200.00INR / Each

Buy It Together

Current Item

Ohaus 30400138 Double Tier Platform Kit, Stainless Steel, 29.8 x 22.2 cm, for use with Waving Shaker SHWV02DG
Ohaus 30400138 Double Tier Platform Kit, Stainless Steel, 29.8 x 22.2 cm, for use with Waving Shaker SHWV02DG
5302755

₹49,200.00INR / Each
+ =
Total Price 66400.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty

More About this Item

Maximize the potential of your rocking and waving shakers with our double-tier platform kits and dimpled mats. The double-tier platforms expand your workspace, allowing you to process more samples simultaneously. Complement these platforms with our dimpled mats, designed to securely hold tubes in place and prevent shifting. This combination not only increases your sample capacity but also enhances efficiency and organization in your laboratory. Upgrade today to optimize your shaker’s performance and streamline your workflow.

1 Year Warranty
Specifications
  • MaterialStainless Steel
  • Compatible WithOhaus Waver SHWV02DG
  • Length (InchShort)11 3/4
  • Width (InchShort)8 3/4
  • Width (cm)22.2
  • Length (cm)29.8
  • DescriptionDouble Tier Platform Kit, Stainless Steel, 29.8 x 22.2 cm, for use with Waving Shaker SHWV02DG
  • Warranty1 year
Reviews
Q&A