Ohaus 30252145 Pipette Adjustment Kit, for Explorer Semi-Micro Balances
Representative image only.
Ohaus 30252145 Pipette Adjustment Kit, for Explorer Semi-Micro Balances
Ohaus | Mfr: 30252145| Item: 2060564
₹613,000.00INR / Each
₹613,000.00INR / Each
Product Description
Includes
Glass plate cover, aluminum trap, wind ring, aluminum reservoir, and carrying case.
Key Features
  • Perform pipette adjustments in-house
  • Install easily without tools
  • Allows for testing of large and small volume pipettes
  • Stores safely in dedicated carrying case
  • 1 year warranty
More About this Item
Perform routine pipette tests and adjustments in the lab efficiently. This pipette adjustment kit lets you verify a pipette’s dosage is within tolerance and significantly reduces the rate of evaporation in comparison to using an open beaker to help ensure reliable results. A 120 mL reservoir handles high volume pipettes or pipettes as low as 100 µL volume. Components come in a portable hard case for secure storage and transport. Compatible with Explorer Plus Semi-Micro, Analytical, and Precision models. Not for use with high-capacity model.
1 Year Warranty
Specifications
  • Compatible WithExplorer and Explorer Plus Analytical, Precision, and Semi-Micro Balances
  • DescriptionPipette Adjustment Kit for Explorer Semi-Micro Ohaus Balances
  • WarrantyOne year