Ohaus 30037451 In Use Cover for Ranger 3000/Ranger Count 3000
Representative image only.
Ohaus 30037451 In Use Cover for Ranger 3000/Ranger Count 3000
Ohaus | Mfr: 30656204| Item: 1161097
Protect your display and controls—even while using your balance
₹15,400.00INR / Each
₹15,400.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Protect your balance from splashes, spills, and dust with these convenient covers. Covers will not affect weighing accuracy.
1 Year Warranty
Specifications
  • LanguagesEnglish
  • Height (in)1
  • Width (in)4
  • Depth (in)6
  • DescriptionIn Use Cover for Ranger 3000/Ranger Count 3000