Ohaus 30037450 WB Hook Kit for Ranger 3000/Ranger Count 3000/Valor 7000
Representative image only.
Ohaus 30037450 WB Hook Kit for Ranger 3000/Ranger Count 3000/Valor 7000
Ohaus | Mfr: 30037450| Item: 1161096
Expand the uses of your balance with the below-balance weighing hook
₹47,000.00INR / Each
₹47,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
A hook is available underneath all balances for weighing beneath the balance. This is particularly useful in applications such as determining the measurment of large pieces.
1 Year Warranty
Specifications
  • LanguagesEnglish
  • Height (in)1
  • Width (in)4
  • Depth (in)6
  • DescriptionWB Hook Kit for Ranger 3000/Ranger Count 3000/Valor 7000