Environmental Express Oakton pH/Ion 700 Ion 700 Benchtop Meter with Probes

Representative image only.

Environmental Express Oakton pH/Ion 700 Ion 700 Benchtop Meter with Probes

Designed for today's crowded laboratory benchtops!

₹122,000.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹122,000.00INR / Each

Buy It Together

Current Item

Environmental Express Oakton pH/Ion 700 Ion 700 Benchtop Meter with Probes
Environmental Express Oakton pH/Ion 700 Ion 700 Benchtop Meter with Probes
3541920

₹122,000.00INR / Each
+ + + + + + +
Environmental Express pH 2000 Benchtop Meter Kit
3541954

₹145,000.00INR / Each
+ =
Total Price 820400.00 ADD 9 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing.

More About this Item

Oakton benchtop 700-series meters are designed for use in crowded laboratory settings. Compact footprint of less that 6" x 7" (16 x 18 cm) conserves valuable space, while the large, dual-display LCD provides excellent visibility—even from a distance.

Ion 700 meter measures pH, mV, relative mV, ion concentration and temperature. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Models feature five-point pH calibration with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers. Meters additionally feature ion concentration calibration of up to 5 points.

Meter features include Auto-Hold, °C measurement, and easy recall of electrode slope or offset. The convenient pull-out quick reference card keeps procedures handy at all times. Use 700-series meters with any pH, ORP, or ISE electrodes with a BNC connector.

3 Year WarrantyConformite Europeenne
Specifications
  • Meter TypeBenchtop
  • Display typeLCD
  • ConnectionspH: BNC; Temperature: 2.5 mm
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0.01 ppm
  • Max ISE1999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.5% of reading
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature compensationAutomatic or manual
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Width (cm)17.4625
  • Height (cm)6.985
  • Width (in)6 7/8
  • Height (in)2 3/4
  • Probe IncludedpH and ATC probes
  • DescriptionIon 700 Benchtop Meter with Probes
Reviews
Q&A