Nylon Bottle/Cylinder Brush, 24" Handle x 6" L x 2.75" Diameter; 12/Pk

Representative image only.

Nylon Bottle/Cylinder Brush, 24" Handle x 6" L x 2.75" Diameter; 12/Pk

| Item: 8455019

₹38,400.00INR / Pkg of 12

₹38,400.00INR / Pkg of 12

Buy It Together

Current Item

Nylon Bottle/Cylinder Brush, 24" Handle x 6" L x 2.75" Diameter; 12/Pk
Nylon Bottle/Cylinder Brush, 24" Handle x 6" L x 2.75" Diameter; 12/Pk
8455019

₹38,400.00INR / Pkg of 12
+ =
Total Price 44200.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 month warranty

More About this Item

Ideal for cleaning bottles, cylinders, and large test tubes. Full radial end tip promotes effective cleaning and reduces scratching of glass and plasticware. Wire handle has hanging hook.

1 Month Warranty
Specifications
  • MaterialNylon
  • ColorBlack
  • Handle Length (in)24
  • DescriptionNylon Bottle/Cylinder Brush, 24" Handle x 6" L x 2.75" Diameter; 12/Pk
  • Warranty1 month
Reviews
Q&A