Nordson Divert 1-Way Stopcock, High Pressure, Polycarbonate, Female Luer x Locking Male Luer; 1/Ea
Representative image only.
Nordson Divert 1-Way Stopcock, High Pressure, Polycarbonate, Female Luer x Locking Male Luer; 1/Ea
| Mfr: FCVHPFMR-001| Item: 5614091
Easily control the flow of a liquid or gas using an on/off diverter
₹980.00INR / Each
₹980.00INR / Each
Product Description
Key Features
  • Diverter arrows indicate flow direction
  • Low-torque handle provides smooth adjustments
  • DEHP free
  • ISO 80369-7 compliant
  • Withstands pressures up to 500 psi
  • Acetal diverter
  • Female Luer x Locking Male Luer
  • 1 each
  • Manufacturer defect warranty
More About this Item
Add a stopcock to your system for an easy on/off control of your fluid. One-way stopcocks feature one female Luer fitting and a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Diverter arrows indicate flow direction, enhancing operational efficiency and minimizing errors. Transparent polycarbonate body makes it easy to see the flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolycarbonate
  • Connection TypeLuer x Luer
  • Number Of Ports2
  • Max Operating Pressure (PSI)500
  • Regulatory ApprovalsNon-DEHP
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Locking Male Luer
  • Quantity1
  • Description1-Way Stopcock, High Pressure, Polycarbonate, Female Luer x Locking Male Luer; 1/Ea
  • WarrantyManufacturer defect