Nordson Divert 1-Way Stopcock, Gamma-Stable Polycarbonate, Female Luer x Male Luer; 25/Pk
Representative image only.
Nordson Divert 1-Way Stopcock, Gamma-Stable Polycarbonate, Female Luer x Male Luer; 25/Pk
| Mfr: FCVFM-001| Item: 5614096
Easily control the flow of a liquid or gas using an on/off diverter
₹8,700.00INR / Pkg of 25
₹8,700.00INR / Pkg of 25
Product Description
Key Features
  • Diverter arrows indicate flow direction
  • Low-torque handle provides smooth adjustments
  • DEHP free
  • ISO 80369-7 compliant
  • Withstands gamma irradiation
  • HDPE diverter
  • Female Luer x Male Luer
  • Pack of 25
  • Manufacturer defect warranty
More About this Item
Add a stopcock to your system for an easy on/off control of your fluid. One-way stopcocks feature one female Luer fitting and a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Diverter arrows indicate flow direction, enhancing operational efficiency and minimizing errors. Transparent polycarbonate body makes it easy to see the flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialGamma-Stable Polycarbonate
  • Sterilization MethodGamma irradiation
  • Connection TypeLuer x Luer
  • Number Of Ports2
  • Max Operating Pressure (PSI)50
  • Regulatory ApprovalsNon-DEHP
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Male Luer
  • Quantity25
  • Description1-Way Stopcock, Gamma-Stable Polycarbonate, Female Luer x Male Luer; 25/Pk
  • WarrantyManufacturer defect